Learn More
CPC Scientific Ac-Tyr-Val-Lys(Biotin)-Asp-2,6-dimethylbenzoyloxymethylketone 1MG
SDP
Supplier:  CPC Scientific CASP014B
Encompass
Inhibitor of caspase-1 that affinity labels active apoptosis-associated caspase-1 related proteases in cell-free extracts and analyzes these proteases preference using cleavage site peptide. ONE-LETTER SEQUENCE: Ac-YVK(Biotin)-D-2,6-diMhylbenzoyloxyMhylketone MOLECULAR FORMULA: C46H63N7O12S1MOLECULAR WEIGHT:938.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [154719-25-2], RESEARCH AREA: ApoptosisREFERENCES: A. Takahashi et al., Proc. Natl. Acad. Sci. USA, 93, 8395 (1996)
Catalog No. 50-210-8947