Learn More
Apexbio Technology LLC Ac-VDVAD-pNA 189684-53-5 5mg
SDP
Supplier:  Apexbio Technology LLC A87565
Encompass_Preferred
Ac-VDVAD-pNA (189684-53-5) is a synthetic peptide substrate designed for monitoring caspase-2 enzymatic activity It is used to detect and quantify the activity of caspase-2 a protease involved in apoptosis Ac-VDVAD-pNA exerts its biological application primarily through chromogenic detection upon cleavage at the VDVAD sequence by activated caspase-2 it releases p-nitroaniline allowing colorimetric measurement of enzyme activity Based on these properties Ac-VDVAD-pNA is employed in apoptosis research and the analysis of caspase-2-mediated cellular processes including studies relevant to myelodysplastic syndromes and cell death signaling pathways
Catalog No. 50-000-95966