Learn More
Apexbio Technology LLC Ac-VEID-AFC 5mg
SDP
Supplier:  Apexbio Technology LLC A87515
Encompass_Preferred
Ac-VEID-AFC is a synthetic fluorogenic substrate targeting caspase-6 It is designed to enable the detection of caspase-6 enzymatic activity thereby facilitating the study of apoptotic pathways Ac-VEID-AFC exerts its biological activity primarily through a cleavage mechanism caspase-6 recognizes and cleaves the VEID peptide sequence releasing the AFC fluorophore which yields a measurable fluorescent signal In in vitro studies Ac-VEID-AFC demonstrates substrate activity enabling quantitative assessment of caspase-6 function Based on these properties Ac-VEID-AFC holds research potential in apoptosis assays and studies investigating caspase-6 activation and downstream apoptotic signaling
Catalog No. 50-000-92971