Learn More
Cayman Chemical Ac-YVAD-AFC 1mg
SDP
Supplier:  Cayman Chemical 175911 MG
Encompass_Preferred
A fluorogenic substrate whose amino acids YVAD have been shown to be a preferred cleavage site for caspase-1 and -4
Catalog No. 50-298-0589