Learn More
Enzo Life Sciences Ac-YVAD-AMC (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260024M001
Encompass_Preferred
Caspase-1 substrate. Fluorogenic caspase-1 substrate with a Km=14µM. Ex.:340-360nm, Em.:440-460nm.Sequence is based on precursor interleukin-1β Asp116 cleavage site for caspase-1. Alternative name: Caspase-1 substrate (fluorogenic). Formula: C33H39N5O10. MW: 665.7. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Peptide content: 75-95%. Solubility: Soluble in DMSO. Dilute with distilled water or buffer, pH 7.0-7.5 (e. g. TRIS, HEPES). Long Term Storage: -20°C. Handling: Protect from light. Keep cool and dry.
Catalog No. 50-200-8230