Learn More
Enzo Life Sciences Ac-YVAD-CHO (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260027M001
Encompass_Preferred
Inhibitor of caspase-1 and -4. Potent and reversible inhibitor of caspase-1. Also inhibits caspase-4. Strongly inhibits anti-Fas induced apoptosis in L929-Fas cells. Alternative name: Caspase-1 inhibitor (aldehyde). Formula: C23H32N4O8. MW: 492.5. Purity: ≥96% (HPLC). Appearance: White powder. Formulation: Lyophilized. Peptide content: 70-90%. Solubility: Soluble in DMSO or water (both 50mg/ml). Long Term Storage: -20°C. Use/Stability: Stock solutions should be divided into aliquots and stored at -20°C. Handling: Keep under inert gas. Keep cool and dry.
Catalog No. 50-200-8234