Learn More
CPC Scientific Ac-Tyr-Val-Ala-Asp-CMK 0.5MG
SDP
Supplier:  CPC Scientific CASP012A
Encompass
Irreversible inhibitor of caspase-1 (ICE) that has also been shown to be a potent therapeutic agent against in vivo liver apoptosis in mice.ONE-LETTER SEQUENCE: Ac-YVAD-CMKMOLECULAR FORMULA: C24H33N4O8Cl1MOLECULAR WEIGHT:541STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [178603-78-6], RESEARCH AREA: ApoptosisREFERENCES: J. B. Schulz et al., J Neurosci, 16, 4696 (1996); N.P.C. Walker et al., Cell 78, 343 (1994)
Catalog No. 50-210-8942