Learn More
Carbosynth LLC. Ac-Tyr-Val-Ala-Asp-CH2Cl [Ac-YVAD-CMK] (5 mg vial), 1ea, MW: 540.99, Molecular formula: C24H33N4O8Cl, CAS# 178603-78-6
SDP
Supplier:  Carbosynth LLC. ICL3180V5MG
Encompass_Preferred
Synonyms: Acetyl-L-Tyrosyl-L-Valyl-L-Alanyl-L-Aspart-1-yl Chloromethylketone, Application: Inhibitor for Caspases, Storage Temperature: -20°C, References: Y.A. Lazebnik, S.H. Kaufmann, S. Desnoyers, G.G. Poirier, and W.C. Earnshaw, Nature, 371, 346 (1994)., M. Enari, H. Hug, and S. Nagata, Nature, 375, 78 (1995)., C.E. Milligan, D. Prevette, H. Yaginuma, S. Homma, C. Cardwell, L.C. Fritz, K.J. Tomaselli, R.W. Oppenheim, and L.M. Schwartz, Neuron, 15, 385 (1995).
Catalog No. 50-168-5841