Learn More
Medchemexpress LLC Ac4GalNAz | 653600-56-7 | >99.9% | C16H22N4O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14112850MG
Encompass_Preferred
Ac4GalNAz is an alkyl chain-based PROTAC linker designed for synthesizing PROTACs. It functions as a click chemistry reagent, enabling selective degradation of target proteins by leveraging the intracellular ubiquitin-proteasome system.
  • Contains an azide group for click chemistry reactions
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • Facilitates PROTAC synthesis
Catalog No. 50-003-54899