Learn More
Selleck Chemical LLC Ac4ManNAz E7558-5MG
SDP
Supplier:  Selleck Chemical LLC E75585MG
Encompass_Preferred
Ac4ManNAz is an azido-containing metabolic glycoprotein labeling reagent Ac4ManNAz can be used to selectively modify proteins Ac4ManNAz can be used in cell labeling tracking and proteomic analysis Ac4ManNAz is a click chemistry reagent it contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-000-41015