Learn More
Medchemexpress LLC Acacetin 7-O-glucuronide | 38226-83-4 | 460.39 | 1 MG

Supplier: Medchemexpress LLC HYN120281MG
Acacetin 7-O-glucuronide is a glucuronide isolated from the methanolic leaf extract of Acacetin. It has potential applications in the development of nutraceuticals and pharmaceutical formulations.
- Isolated from methanolic leaf extract of Acacetin.
- Potential applications in the development of nutraceuticals and pharmaceutical formulations.
- Purity: 99.30%
- Appearance: Solid
- Color: Off-white to light yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.