Learn More
Selleck Chemical LLC AICAR (Acadesine) 200mg 2627-69-2 NSC105823
SDP
Supplier:  Selleck Chemical LLC S1802200MG
Encompass_Preferred
AICAR (Acadesine, NSC105823), an AMPK activator, results in accumulation of ZMP, which mimics the stimulating effect of AMP on AMPK and AMPK kinase. AICAR (Acadesine) induces mitophagy. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-792-6