Learn More
Medchemexpress LLC Acalabrutinib-d4 | 2699608-18-7 | 99.5% | 469.53 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17600S5MG
Encompass_Preferred
Acalabrutinib-d4 (ACP-196-d4) is a deuterium labeled Acalabrutinib. Acalabrutinib (ACP-196) is an orally active, irreversible, and highly selective second-generation BTK inhibitor. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Deuterium labeled Acalabrutinib
  • Orally active, irreversible, and highly selective second-generation BTK inhibitor
  • Click chemistry reagent containing an Alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-87333