Learn More
Medchemexpress LLC Acalisib | 870281-34-8 | 100.0% | 401.40 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1264450MG
Encompass_Preferred
Acalisib is a potent and selective PI3Kδ inhibitor with an IC50 of 12.7 nM. It demonstrates greater selectivity for PI3Kδ compared to other PI3K class I enzymes and related kinases. This product is intended for research use only and not for sale to patients.
  • Potent and selective PI3Kδ inhibitor
  • High selectivity against other PI3K class I enzymes and related kinases
  • Demonstrates IC50 of 12.7 nM for PI3Kδ
  • Useful in biochemical in vitro lipid kinase assays
  • Applicable in cell survival evaluation via MTT assay
  • Proven to reduce multiple myeloma xenograft growth in in vivo studies
Catalog No. 50-004-38197