Learn More
Medchemexpress LLC Acalisib | 870281-34-8 | 99.98% | 401.40 | 100MG
SDP
Supplier:  Medchemexpress LLC HY12644100MG
Encompass_Preferred
Acalisib is a potent and selective PI3Kδ inhibitor with an IC50 of 12.7 nM. It is for research use only and not sold to patients.
  • More selective for PI3Kδ (IC50=12.7 nM) compared to other PI3K class I enzymes.
  • Reduces PDGF-induced pAkt by 50% at 11,585 nM and LPA-induced pAkt by 50% at 2,069 nM in fibroblasts.
  • Sufficient to reduce the growth of multiple myeloma xenografts in mice.
Catalog No. 50-004-38284