Learn More
Medchemexpress LLC Acalisib | 870281-34-8 | 99.98% | 401.40 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY12644
Encompass_Preferred
Acalisib is a potent and selective PI3Kδ inhibitor with an IC50 of 12.7 nM. It demonstrates high selectivity against other PI3K class I enzymes and related kinases, making it suitable for targeted research. Studies have shown its effectiveness in reducing the growth of multiple myeloma xenografts in mice. This product is for research use only.
  • Potent and selective PI3Kδ inhibitor
  • IC50 of 12.7 nM against PI3Kδ
  • High selectivity over other PI3K class I enzymes
  • Reduces growth of multiple myeloma xenografts in mice
  • For research use only
Catalog No. 50-003-64103