Learn More
Selleck Chemical LLC Acamprosate Calcium S3748-25mg
SDP
Supplier:  Selleck Chemical LLC S374825MG
Encompass_Preferred
Acamprosate (N-Acetylhomotaurine Calcium acetylhomotaurinate N-acetylhomotaurinate) is a synthetic compound with a chemical structure similar to the amino acid neurotransmitter gamma-aminobutyric acid (GABA) and the amino acid neuromodulator taurine
Catalog No. 50-310-9186