Learn More
Medchemexpress LLC Acamprosate (calcium) | 77337-73-6 | 99.9% | 200.24 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1703010MM/1ML
Encompass_Preferred
Acamprosate calcium (Campral EC) acts as a GABA receptor agonist and modulator of glutamatergic systems, reducing alcohol consumption in animal models of alcohol addiction. It inhibits Ca2+ influx through NMDA receptors and voltage-dependent Ca2+ channels.
  • Inhibits Ca²⁺ influx through both NMDA receptors and voltage-dependent Ca²⁺ channels.
  • Exhibits antidepressant-like effects in mouse tail suspension test models.
  • Ameliorates alcohol-induced cognitive deficits and shows anti-relapse effects in mouse models.
Catalog No. 50-003-84209