Learn More
Medchemexpress LLC Acanthopanaxoside B | 915792-03-9 | 25 MG

Supplier: Medchemexpress LLC 25MGHYN4135
Acanthopanaxoside B is a triterpenoid saponin isolated from the leaves of *Acanthopanax senticosus*, primarily used for scientific research as a natural product.
- Molecular formula: C₆₁H₉₈O₂₇
- Molecular weight: 1263.41
- General storage: 4°C, protect from light
- In solvent storage: -80°C for 6 months; -20°C for 1 month (protect from light)
- Appearance: Solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.