Learn More
Selleck Chemical LLC Acarbose S1271-50mg
SDP
Supplier:  Selleck Chemical LLC S127150MG
Encompass_Preferred
Acarbose(BAY g 5421 Prandase Precose Glucobay Bay-g 5421) is an inhibitor of intestinal alpha-glucosidase used to treat type 2 diabetes mellitus
Catalog No. 50-311-0976