Learn More
Medchemexpress LLC Acarbose | 56180-94-0 | 99.97% | C25H43NO18 | 1 G
SDP
Supplier:  Medchemexpress LLC HYB00891G
Encompass_Preferred
Acarbose (BAY g 5421) is an orally active alpha-glucosidase inhibitor, acting as an antihyperglycemic agent with an IC50 of 11 nM. It can potentiate the hypoglycemic effects of sulfonylureas or insulin. It is suitable for various research applications.
  • Orally active alpha-glucosidase inhibitor
  • Potentiates hypoglycemic effects of sulfonylureas or insulin
  • Decreases fasting blood glucose
  • Regulates glucose tolerance
  • Suppresses serum IL6 and TNF-α
  • Decreases neointimal Ras and β-galactosidase expression
Catalog No. 50-004-33416