Learn More
Medchemexpress LLC Acarbose (Standard) | 56180-94-0 | 99.9% | C25H43NO18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0089R
Encompass_Preferred
Acarbose (Standard) is an analytical standard of Acarbose, intended for research and analytical applications. This antihyperglycemic agent is an orally active alpha-glucosidase inhibitor with an IC50 of 11 nM, capable of enhancing the hypoglycemic effects of sulfonylureas or insulin.
  • Serves as a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
  • Has a molecular weight of 645.60
  • Appears as a white to off-white solid
  • Initial source is microorganisms
  • Structure classified under saccharides and polysaccharides
  • Shipped at room temperature in the continental US; conditions may vary elsewhere
Catalog No. 50-003-57997