Learn More
Medchemexpress LLC Acarbose (standard) | 56180-94-0 | 99.9% | C25H43NO18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0089R50MG
Encompass_Preferred
Acarbose (Standard) is an analytical standard of Acarbose, intended for research and analytical applications. This orally active alpha-glucosidase inhibitor (IC50=11 nM) can potentiate the hypoglycemic effects of sulfonylureas or insulin. It is an analytical standard grade suitable for qualitative, quantitative, and methodological research experiments.
  • Analytical standard for Acarbose
  • Intended for research and analytical applications
  • Orally active alpha-glucosidase inhibitor (IC50=11 nM)
  • Can potentiate hypoglycemic effects of sulfonylureas or insulin
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-32046