Learn More
Waters Corp Accell Plus CM PrepPak Column, 40 µm, 47 mm x 300 mm, 1/pk
Supplier:  Waters Corp WAT036545
Encompass_Preferred
Waters AccellPlus CM cation exchange packing materials are polymer-coated, silica-based media of 37-55 μm particles with a 300Å pore size for protein purifications.
Catalog No. 50-785-384