Learn More
Waters Corp AccellPlus QMA 500 g WAT010741
Supplier:  Waters Corp WAT010741
Encompass_Preferred
Accell Plus ion exchange packings are 40m, 300 angstrom polymer-coated silica-based materials and are available in strong anion and weak cation exchangers. These packings, for all preparative isolations based on ionic interactions, particularly proteins, enzymes and immunoglobulins and be packed into our Advanced Purification (AP) glass columns.
Catalog No. 50-784-984