Learn More
Cytiva Spare Parts and Accessories for TFF devices - Accessory kit consists of 1.8 m (6 ft.) of PharMed* #24 feed/retentate tubing, 0.6 m (24 in.) of 4.8 mm (3/16 in.) Tygon* filtrate tubing, (8) stainless steel tubing clamps, and (1) screw clamp (1/pkg)
Supplier:  Cytiva FS002X70
Encompass
Simplify Processing of 100 mL to 5 L Volumes When Using the Ultrasette™ Device-Self-contained siphoning units allow continuous buffer exchange-Uses less liquid with less handling and mess than conventional dialysis systems-Constructed of clear acrylic with easy-to-read volume graduations-Connections located in base plate for easy access and minimal sample hold-up-Feed pressure gauge monitors sample processing-Three-way sampling valve allows maximum sample recovery without decanting-Peristaltic pump action minimizes shear of sensitive molecules, eliminates air in the system, and reduces foaming-A variable speed controller allows precise adjustment of crossflow during sample processing This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3456