Learn More
Waters Corp AccQ-Tag Column, 60Å, 4 µm, 3.9 mm X 150 mm, 1/pk
Supplier:  Waters Corp WAT052885
Encompass_Preferred
Amino acid analysis is afundamental process in any protein research. Waters developed a new reagent specific for amino acid derivitization that provides enhanced quantitative accuracy and subpicomole detection for hydrolysate amino acids. We offer a system package consisting of an optimized instrument, prepackaged reagents and extensive documentation.
Catalog No. 50-785-690