Learn More
Andwin Scientific ACCUEDGE SCALPEL HANDLE EA
SDP
Supplier:  Andwin Scientific 6256601
Encompass_Preferred
The The Tissue-Tek Accu-Edge Dissecting Scalpels features a pointed blade that are 0.2 millimeters thicker than other brands, which eliminate the risk of bending under cutting pressure. The blades provide consistent and accurate cutting. This product can be easily attached onto a sturdy, chemical- and heat-resistant scalpel handle.
Catalog No. 50-727-459
 Restricted Item