Learn More
ACCUTEC BLADES INC (3P) AccuForge GEM Single Edge Blade Clamshell. SKU# AGBL-7021-0000 .009" Stainless Steel 3-Facet Blades, Aluminum Back, 100 Blades per Clamshell, 50 Clamshells per Case
Supplier:  ACCUTEC BLADES INC (3P) AGBL70210000
Encompass_Preferred
AccuForge Single Edge blades feature .009 stainless steel and 3-facet edges for sharpness. Each clamshell contains 100 individually wrapped blades. Units per Case: 50
Catalog No. NC3721105