Learn More
ACCUTEC BLADES INC (3P) AccuForge GEM Single Edge Blade Dispenser SKU# AGBL-7024-0000 .009" Stainless Steel 3-Facet Degreased & Washed Blades, Stainless Steel Back, 10 Blades per Dispenser, 200 Dispensers per Case
Supplier:  ACCUTEC BLADES INC (3P) AGBL70240000
Encompass_Preferred
AccuForge Single Edge blades feature .009 stainless steel and 3-facet edges for sharpness. Stainless steel backing material provides optimal corrosion resistance. Blades are degreased and washed for least amount of residual oil. Each dispenser holds 10 blades and includes used blade storage. Bulk pack of 200 dispensers per case. Units per case: 200
Catalog No. NC3721117