Learn More
ACCUTEC BLADES INC (3P) AccuForge GEM3 Single Edge Blade Cartridge, with Microcoat. SKU# AGBL-7013-0000 .009" Stainless Steel 3-Facet MicroCoat Degreased Blades, Steel Back, 100 Blades per Cartridge, 10 Cartridges per Case
Supplier:  ACCUTEC BLADES INC (3P) AGBL70130000
Encompass_Preferred
AccuForge Single Edge blades feature .009 stainless steel and 3-facet edges for sharpness. Blades are degreased to remove residual oil. Our proprietary MicroCoat Enrichment Technology reduces cut force and helps the blade maintain a sharp effective edge over multiple uses. Each cartridge contains 100 blades. Fits Gem Runner™ Dispenser AGTL-7000-0000. Made in the USA
Catalog No. NC3721104