Learn More
LAMDA BIOTECH INC AccuRuler Prestain Prot Ladder
Supplier:  LAMDA BIOTECH INC G02101
Encompass_Preferred
A pre-stained protein molecular weight marker containing 10 proteins ranging from 10 to 180 kDa, formulated for protein electrophoresis and Western blot applications
Catalog No. NC3724881