Learn More
Seracare Life Sciences Inc Anti-SARS-CoV-2 Reference Material Kit - Series 2000, 4 x 3 mL
Supplier:  Seracare Life Sciences Inc 20150225
Encompass
Offers positive materials for antibodies to the SARS-CoV-2 virus. Also included are complementary negative materials. The kit is designed to support assay installation and monitoring of day-to-day assay performance, providing a complete quality solution for SARS-CoV-2 antibody testing.
Catalog No. NC1826191