Learn More
Supply Solutions ACCUSPIN™ Tubes Sterile, 50 mL Capacity, 10ea
Supplier:  Supply Solutions SIGMA A205510EA
Encompass_Preferred
ACCUSPIN™ Tubes are intended for use with the density gradient separation medium Histopaque -1077 in the isolation of lymphocytes and other mononuclear cells. Polypropylene radiation sterilized tube fitted with a high density polyethylene barrier. Each tube will accept 15 mL of the density gradient.
Catalog No. 50-161-3334