Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AccuStandard, Inc AccuStandard Diallyl phthalate Certified Reference Material, 1 component, 100 mg, CAS 131-17-9
SDP

Supplier:  AccuStandard, Inc J002

Encompass

AccuStandard Diallyl phthalate is a Certified Reference Material (CRM), ideal for analytical applications requiring a reliable single-component standard. This product features CAS number 131-17-9 and a molecular formula of C14H14O4, supplied as 100 mg. Store at ambient temperatures above 5°C.

  • Certified Reference Material (CRM) for analytical accuracy.
  • Single-component solution.
  • Storage condition: Ambient (>5°C).
  • Molecular formula: C14H14O4.
  • Molecular weight: 246.26.
  • Melting point: -70°C / -94°F.
  • Boiling point: 165°C to 167°C / 329°F to 333°F.
  • Flash point: 166°C / 331°F - closed cup.
  • Unit: 100 mg.

Catalog No. NC3991854


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.