Learn More
Mac-Mod Avantor ACE C18-PFP, 100 A, 3 um, 2.1 x 150 mm HPLC Column
SDP
Supplier:  Mac-Mod ACE11101502
Encompass
Avantor ACE HPLC columns are manufactured with 100 A Ultra-Inert Base Deactivated Silica specifically designed for your HPLC method development needs. Avantor ACE HPLC columns are available in a wide range of bonded phases (including C18, C18-AR, C18-PFP, C18-HL, C8, C4, Phenyl, AQ, CN and Sil) - Note C18-HL is 95 A. Avantor ACE HPLC columns are available in 3, 5 and 10 um particle sizes and a wide range of column IDs and column lengths including nano, capillary, microbore, analytical, semi-preparative and preparative formats. All Avantor ACE columns are known to deliver excellent peak shape and verified batch to batch reproducibility. Columns are packed in 275 bar HPLC hardware. Note: Avantor ACE HPLC guard columns are available for all 3.0 and 5.0 um particle sizes across 2.1 mm to 4.6 mm column ID - Note: C0001 male/male union is needed to connect guard holder (H0001) to the analytical column. Avantor ACE UHPLC alternatives columns are available under the ACE Excel brand.
Catalog No. 50-752-8057