Learn More
Genetex Inc ACE2 Monoclonal Antibody [MSVA-919R] HistoMAX™ (GTX04425) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04425
Encompass_Preferred
The ACE2 antibody [MSVA-919R] HistoMAX™ (GTX04425) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human clear cell renal cell carcinoma (ccRCC) tissue using GTX04425 ACE2 antibody [MSVA-919R] HistoMAX. Clear cell renal cell carcinoma strong strong membraous ACE2 positivity.
Catalog No. NC3666923