Learn More
Medchemexpress LLC ACE2 Human HEK293 10ug
SDP
Supplier:  Medchemexpress LLC HYP7348910UG
Encompass_Preferred
The ACE2 protein is an important regulator of blood volume and cardiovascular homeostasis ACE2 Protein Human (HEK293 His) is the recombinant human-derived ACE2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80072