Learn More
Medchemexpress LLC Acebutolol (hydrochloride) | 34381-68-5 | 372.89 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY17497A10MM/1ML
Encompass_Preferred
Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. It is used in the treatment of hypertension, angina pectoris, and cardiac arrhythmias.
  • Orally active β1 adrenergic receptor (β1AR) antagonist
  • Used in the treatment of hypertension
  • Used in the treatment of angina pectoris
  • Used in the treatment of cardiac arrhythmias
  • Targets adrenergic receptor
  • Inhibits β1 adrenoceptor
Catalog No. 50-003-86777