Learn More
Medchemexpress LLC Acebutolol (hydrochloride) | 34381-68-5 | 99.7% | 372.89 | 1 G
SDP
Supplier:  Medchemexpress LLC HY17497A1G
Encompass_Preferred
Acebutolol hydrochloride is an orally active β1 adrenergic receptor (β1AR) antagonist. It is used in the treatment of hypertension, angina pectoris, and cardiac arrhythmias. It acts as a beta blocker and has been shown to decrease cardiac output and significantly reduce regional blood flow in in vivo studies.
  • Orally active β1 adrenergic receptor (β1AR) antagonist
  • Used to treat hypertension
  • Used to treat angina pectoris
  • Used to treat cardiac arrhythmias
  • Beta blocker for hypertension and arrhythmias
  • Decreases cardiac output in in vivo studies
  • Reduces regional blood flow in in vivo studies
Catalog No. 50-003-87182