Learn More
Medchemexpress LLC Aceclidine (quinuclidin-3-yl acetate) | 827-61-2 | 98.0% | 169.22 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3206710G
Encompass_Preferred
Aceclidine (Quinuclidin-3-yl acetate) is a modulator of the M3 muscarinic acetylcholine receptor and an M1 receptor agonist (EC50: 40 μM). It acts as a cycloplegic agent, a surfactant, a tonicity adjustor, and optionally a viscosity enhancer and an antioxidant. This compound holds potential for research into various disorders, including refractive errors of the eye, xerostomia, Sjogren's syndrome, glaucoma, conjunctivitis, lacrimal gland disease, and esotropia.
  • Modulates M3 muscarinic acetylcholine receptor
  • Acts as an M1 receptor agonist
  • Functions as a cycloplegic agent
  • Acts as a surfactant and tonicity adjustor
  • Can act as a viscosity enhancer and antioxidant
  • Useful for research into eye disorders like glaucoma and refractive errors
  • Relevant for studies on xerostomia and Sjogren's syndrome
Catalog No. 50-004-00439