Learn More
Medchemexpress LLC HY-B0634 500mg Medchemexpress, Aceclofenac CAS:89796-99-6 Purity:>98%
Supplier: Medchemexpress LLC HYB0634500MG
Medchemexpress, HY-B0634 500mg Aceclofenac CAS:89796-99-6 Aceclofenac is an orally active nonsteroidal anti-inflammatory drug (NSAID), with analgesic and anti-inflammatory properties. Aceclofenac is used for the research of osteoarthritis, ankylosing spondylitis, rheumatoid arthritis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.