Learn More
Medchemexpress LLC Acedoben | 556-08-1 | 99.9% | 10 G
SDP
Supplier:  Medchemexpress LLC HYB125010G
Encompass_Preferred
Acedoben is a biochemical agent. When combined with iron ions, it forms a fast self-assembled coordination complex called Fe-Ace. This complex serves as a carrier for tumor antigens and enhances antigen-specific anti-tumor immunity due to its inherent adjuvant properties. Acedoben is for research use only and not sold to patients.
  • Serves as a carrier of tumor antigens.
  • Enhances antigen-specific anti-tumor immunity due to its inherent adjuvant properties.
Catalog No. 50-004-12874