Learn More
Medchemexpress LLC Acedoben | 556-08-1 | >99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1250R100MG
Encompass_Preferred
Acedoben (Standard) is an analytical standard and biochemical reagent from MedChemExpress, intended for research and analytical applications. Acedoben is a biochemical agent that, in coordination with iron ions, can construct a fast self-assembled coordination complex. This Fe-Ace complex can function as a carrier for tumor antigens and enhance antigen-specific anti-tumor immunity due to its inherent adjuvant properties.
Features and Benefits:
  • Analytical standard for Acedoben research
  • Biochemical reagent for various applications
  • Forms self-assembled coordination complexes with iron ions
  • Potential for enhancing anti-tumor immunity
Catalog No. 50-004-12813