Learn More
Selleck Chemical LLC Acemetacin S2602-50mg
SDP
Supplier:  Selleck Chemical LLC S260250MG
Encompass_Preferred
Acemetacin (K-708 TVX 1322) is a non-steroidal anti-inflammatory drug and a glycolic acid ester of indometacin that is a cyclooxygenase inhibitor
Catalog No. 50-311-2525