Learn More
Apexbio Technology LLC Acemetacin 53164-05-9 5g
Supplier: Apexbio Technology LLC B14405000
Acemetacin (CAS 53164-05-9) is a non-steroidal anti-inflammatory compound structurally classified as a glycolic acid ester derivative of indometacin It exerts its pharmacological effects by inhibiting cyclooxygenase (COX) enzymes thereby suppressing prostaglandin synthesis involved in inflammation and pain signaling pathways Acemetacin is commonly employed in biomedical research to investigate COX-mediated processes and the modulation of inflammatory responses as well as to explore pathways related to non-steroidal anti-inflammatory drug (NSAID) pharmacodynamics and safety profiles