Learn More
Medchemexpress LLC Acenaphthylene-d8 | 93951-97-4 | 99.5% | 160.24 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW013570S100MG
Encompass_Preferred
Acenaphthylene-d8 is the deuterium labeled Acenaphthylene. Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH). PAHs are derived naturally from coal and tar deposits, and produced by incomplete combustion of organic matter.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54483