Learn More
Medchemexpress LLC Acenaphthylene-d8 | 93951-97-4 | 99.5% | 160.24 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW013570S50MG
Encompass_Preferred
Acenaphthylene-d8 is the deuterium labeled Acenaphthylene. Acenaphthylene is a polycyclic aromatic hydrocarbon (PAH) derived naturally from coal and tar deposits and produced by incomplete combustion of organic matter. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-54467