Learn More
Medchemexpress LLC Acephate | 30560-19-1 | MFCD00055361 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0841100MG
Encompass_Preferred
Acephate is a broad-spectrum anticholinesterase insecticide that acts by inhibiting AChE activity of insects. It is used for controlling several species of insects in agriculture and horticulture.
  • Broad-spectrum anticholinesterase insecticide
  • Inhibits AChE activity in insects
  • Used for control of various insect species in agriculture and horticulture
  • For research use only
Catalog No. 50-004-07608