Learn More
Medchemexpress LLC Acesulfame (potassium) | 55589-62-3 | 25 G
Supplier: Medchemexpress LLC 25GHYD0195
Acesulfame potassium is a synthetic sweetener. It can affect cognitive function by altering neurometabolic functions. It suppresses autophagic degradation of PD-L1 through the ERK1/2-mTORC1-ULK1 pathway, which may be related to immune evasion in cancer cells. It is useful in research on neurological diseases, metabolic disorders, cancer, and immune evasion.
- Synthetic sweetener
- Affects cognitive function by altering neurometabolic functions in mice
- Suppresses autophagic degradation of PD-L1 through the ERK1/2-mTORC1-ULK1 pathway
- Useful in research on neurological diseases, metabolic disorders, cancer, and immune evasion